Learn More
DWK Life Sciences Kimble™ Disposable Soda Lime Pasteur Pipette
The Kimble™ Disposable Soda Lime Pasteur Pipette is manufactured from 90 expansion soda lime glass conforming to USP Type III requirements.
Catalog Number | Mfr. No. | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Price | Quantity & Availability | |||||
22-037-541
|
DWK Life Sciences
63A54 |
14.61 cm |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||||
22-037-544
|
DWK Life Sciences
63B93 |
22.86 cm |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||||
22-037-513
|
DWK Life Sciences
63C50 |
14.61 cm |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View Alternate Product |
N/A | |||||
Description
- Soda lime pipets are manufactured from 90 expansion soda lime glass tubing conforming to USP Type III requirements
- Ideal for the one-time use for small volume liquid transfer
- Available either unplugged or with a cotton plug to suit most applications
Specifications
Soda Lime Glass | |
Non-sterile | |
Single-use |