Learn More
PBL Assay Science VeriKine Human Interferon Alpha (IFN-Alpha, IFNa) ELISA, Multi-Subtype, Tissue Culture Media (TCM) [1x96-well plate]
Supplier: PBL Assay Science 411051
VeriKine Human Interferon Alpha Multi-Subtype ELISA Kit is manufactured by PBL Assay Science (Catalog #41105-1) for research use only (RUO) and packaged as a 1x96-well plate. This IFNa Multi-Subtype ELISA is Tissue Culture Media (TCM) compatible, quantifies 11 of 12 human IFN-Alpha subtypes, and has an assay range of 12.5-500 pg/mL or 156-5,000 pg/mL. This ELISA kit has intra- and inter-assay CVs of less than 10%.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.