Learn More
United States Biological Corporation E2205 EDINBURGH MIN MEDIA 500G
Supplier: United States Biological Corporation E2205500G
Edinburgh Minimal Media is a thiamine-free formulation for nmt1 promoter studies for Schizosaccharomyces pombe. EMM2 is used for vegetative growth.