Learn More
NSI Environmental Solutions COMPLEX NUTRIENTS 2 500ML
Supplier: NSI Environmental Solutions QCI064
Complex Nutrient, Whole Volume Standard for Determination of : TKN: 0.35 to 35mg/L, Total Phosphorus: 0.15 to 10mg/L, 500mL