Learn More
Quadrex Corporation CLMN-PHASE 007-1,5M L,ID 0.53
SDP
Supplier:  Quadrex Corporation 00715V5.0F
Encompass_Preferred
fused silica nonpolar capillary column - phase 007-1, length 5 meters, ID 0.53 mm, Film 5.0 microns.Bonded and CrosslinkedNon polarExcellent thermal stabilityComparable to: SE-30, OV-1, OV-101Similar phases: DB-1, Rtx -1, HP-1, Ultra-1, SPB-1, CPSil-5CB, AT-1, BP-1, DB-Petro, DB-2887, RTx-PONA, RTx-2887, HP-PONA, Petrocol-DH, Petrocol-2887, CP-SimDist-CB, At-PetroUSP Phase Specification: G1, G2
Catalog No. NC1564356