Learn More
Electron Microscopy Sciences Capsule Mold Type B, 12 Cavities
Supplier: Electron Microscopy Sciences 70160
Our molds are made from the strongest possible silicone rubber providing extremely high tear resistance. The molds are are reusable. Type B has 12 cavities, 8mm diameter body, 5mm diameter tip 16mm height, tapered tip.