Learn More
Interscience Lab Inc BAGMIXER 400CC
Supplier:  Interscience Lab Inc 024230
Encompass_Preferred
Request Bulk Order
BagMixer��400 CC��is an easy-to-use and powerful lab blender. Adapted to all kinds of applications and with a guarantee of optimal bacterial extraction, it is a great tool for lab analyses avoiding all risks of cross-contamination.Exclusive Click & Clean� removable paddles (Patented) - Fully opening door (270�) - Security drip tray - All stainless steel 3. USER-FRIENDLY - Compact - Multifunctional screen: easy control - Side-by-side paddle stop: easy insertion of the bag - Q-Tight� closing: no more leaks upwards 4. ACCURACY - Adjustable blending speed (4 - 6 - 8 - 10 strokes per second) - Adjustable blending time (1 second - 1 hour or open-running) - Adjustable blending power : adapted to any kind of sample (Patented)
Catalog No. NC0722912