Learn More
Bioassay Systems EnzyChrom™ Maltose Assay Kit.Quantitative determination of maltose in various biological samples such as serum, urine, food and beverages, etc.
Supplier: Bioassay Systems EMLT100
For quantitative determination of maltose and evaluation of drug effects on its metabolism. Key Features: Fast and sensitive. Use of 10 µL sample. Linear detection range 2 to 500 µM maltose for colorimetric assays and 1 to 50 µM for flourimetric assays. Convenient. The procedure involves adding a single working reagent, and reading the absorbance after 60 minutes. Room temperature assay. No 37°C heater is needed. High-throughput. "Add-mix-read" type assay. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD570nm, or FL530/585nm. Samples: Serum, urine, food and beverages, etc. Species: All. Procedure: Assay takes 60 min. Kit size: 100 tests. Detection limit: 2 µM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.