Learn More
Bioassay Systems EnzyChrom™ Xanthine Oxidase Assay Kit. For quantitative determination of xanthine oxidase enzyme activity.
Supplier: Bioassay Systems EXOX100
For quantitative determination of xanthine oxidase enzyme activity. Key Features: Sensitive and accurate. Use as little as 10 µL samples. Linear detection range in 96-well plate for 20 minute incubation: 0.03 to 25 U/L xanthine oxidase for colorimetric assays and 0.01 to 2.5 U/L for fluorimetric assays. Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 20 min at room temperature. Method: OD570nm, FL530/585nm; Samples: Cell lysate, serum, and other biological samples; Size: 100 tests.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.