Learn More
Bioassay Systems EnzyChrom™ Fumarate Assay Kit. For quantitative determination of fumarate and drug effects on its metabolism.
Supplier: Bioassay Systems EFUM100
For quantitative determination of fumarate and drug effects on its metabolism. Key Features: Fast and sensitive. Use of 20 µL sample. Linear detection range 0.005 to 2 mM fumarate in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the optical density after 30 minutes. Room temperature assay. No 37°C heater is needed. High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD565nm. Samples: Food, beverage and other biological samples (e.g. cell lysate, tissue homogenate, serum). Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 5 µM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.