Learn More
Bioassay Systems EnzyFluo™ Tryptophan Assay Kit. Direct determination of tryptophan in serum.
Supplier: Bioassay Systems ETRP100
For direct determination of tryptophan in serum. Key Features: Fast and sensitive. Use of 10 µL sample. Linear detection range from 10 µM to 400 µM tryptophan in 96-well plate assay. Convenient. The procedure involves adding a single working reagent, and reading the fluorescence after 30 minutes. Room temperature assay. No 37 °C heater is needed. High-throughput. "Add-mix-read" type assay. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: FL530/585nm; Samples: Serum; Size: 100 wells.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.