Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zinc Oxide, puriss. p.a., ACS Reagent, ≥99% (KT), Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Melting Point | ∼1975°C |
Color | White |
pH | ∼7 (at 20°C) |
Quantity | 500 g |
Linear Formula | ZnO |
UN Number | UN3077 |
Formula Weight | 81.39g/mol |
Percent Purity | ≥99% (KT) |
Packaging | Poly Bottle |
Appearance | Powder |
Show More |
Safety and Handling
Hazard Statement | P273-P501 |
DOTInformation | 9/PGIII |
RTECSNumber | 215-222-5 |
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction