Learn More
Med Vet International Boston Round Yorker Dropper Bottle, 30mL/1 oz., 100/Case
Supplier: Med Vet International BSRD30CS
Removable dropper tips allow refilling. Bottle and tips are made of low density polythelene. Screwcap made of polystyrene.