Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Wooden Friction Block with Hook
SCI_ED

Wooden friction blocks with hooks are useful for teaching the basics of force and motion in a simple, yet effective way. A friction block used in tandem with a spring scale is a powerful tool for teaching students about friction, kinetic and potential energy, acceleration, and force

$7.50 - $8.00

Specifications

For Use With (Application) Measuring Force and Mass
For Use With (Equipment) Friction Track
Includes C-Hook
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
S27645
SCI_ED
View Documents
Eisco™
PH0201605F
Wood/Felt
Each for $8.00
Add to Cart
 
S27646
SCI_ED
View Documents
Eisco™
PH0201605S
Wood/Sandpaper
Each for $7.50
Add to Cart
 
Description

Description

  • Wooden friction block with c-hook attached at one end
  • Use with a spring scale to measure force and mass
  • Features 1 wooden side and 1 side covered with a unique material for comparative study
Specifications

Specifications

Measuring Force and Mass
Friction Track
C-Hook
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.