Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Vibration Generator

This Vibration Generator can be used free standing or clamped, and works similar to a loud speaker.
Supplier: Eisco™ VBGN
Description
- Includes a coil wound on a thin tube that moves within a permanent magnetic field when current of an altering nature is applied
- The frequency response encompasses the whole of the audio spectrum and beyond
- Electrical input is made via two 4 mm sockets
- Mechanical output is provided by a shaft
- The unit gives mechanical oscillations when fed by signals from an oscillator / A.F. amplifier or Power Signal
- Maximum Peak to Peak Displacement: 7 to 8 mm at 1 Hz, decreasing with increase in frequency
- Total Frequency Range: 1 to 5 KHz
- Coil Impedance: 3 Ω at 50 Hz
Specifications
Generating Vibration When Current of an Altering Nature is Applied | |
2.4 lb. | |
Vibration Generator |
1 to 5 KHz | |
Oscillator, A.F. Amplifier, or Power Signal Generator |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title