Learn More
TWD Scientific LLC BioX 1D Barcoded 1.0mL Cryovial, Yellow Cap, External Thread. Case of 1000
Supplier: TWD Scientific LLC BX886104
BioX 1.0mL clear polypropylene sterile pre-set 1D barcoded cryovials with external thread and yellow caps assembled. Cryovials have writing patch and marked graduations. 25 cryovials/bag, 20 bags/pack, 2 packs/case, price/case
This item is not returnable.
View return policy