Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ VapLock™ Solvent Delivery Caps

VapLock™ Solvent Delivery Caps reduce chemical evaporation and exposure to hazardous vapors

$56.75 - $124.00

Specifications

For Use With (Application) Solvent Safety
For Use With (Equipment) Solvent-Compatible Containers
Thread Size (English) 1/4 in.-28 UNF(F)
View More Specs

Products 31
Catalog Number Mfr. No. Type Material Size Price Quantity & Availability  
Catalog Number Mfr. No. Type Material Size Price Quantity & Availability  
13-311-003
View Documents
Cole-Parmer™
1201803
Two 1/4 in.-28 ports Blue/white PTFE, PP GL45
Each for $71.25
Add to Cart
 
13-311-025
View Documents
Cole-Parmer™
1201825
304 SS Port Thread Inserts, three 1/4 in.-28 White PTFE, PP, SS Nalgene 38-430
Each for $81.00
Add to Cart
 
13-311-037
View Documents
Cole-Parmer™
1201837
304 SS Port Thread Inserts, three 1/4 in.-28 White PTFE, PP, SS Wako 38 mm
Each for $77.00
Add to Cart
 
13-311-052
View Documents
Cole-Parmer™
1201852
Three 1/4 in.-28 ports Blue/white PTFE, PP GL32
Each for $65.75
Add to Cart
 
13-311-004
View Documents
Cole-Parmer™
1201804
Three 1/4 in.-28 ports Blue/white PTFE, PP GL45
Each for $71.50
Add to Cart
 
13-311-050
View Documents
Cole-Parmer™
1201850
304 SS Port Thread Inserts, four 1/4 in.-28 Blue/white PTFE, PP, SS GL32
Each for $92.00
Add to Cart
 
13-311-028
View Documents
Cole-Parmer™
1201828
Three 1/4 in.-28 ports White PTFE, PP Nalgene 38-430
Each for $69.25
Add to Cart
 
13-311-013
View Documents
Cole-Parmer™
1201813
304 SS Port Thread Inserts, three 1/4 in.-28 Black/white PTFE, PP, SS GL38
Each for $80.00
Add to Cart
 
13-311-015
View Documents
Cole-Parmer™
1201815
Two 1/4 in.-28 ports Black/white PTFE, PP GL38
Each for $68.25
Add to Cart
 
13-311-051
View Documents
Cole-Parmer™
1201851
Two 1/4 in.-28 ports Blue/white PTFE, PP GL32
Each for $76.00
Add to Cart
 
13-311-005
View Documents
Cole-Parmer™
1201805
Four 1/4 in.-28 ports Blue/white PTFE, PP GL45
Each for $77.00
Add to Cart
 
13-311-024
View Documents
Cole-Parmer™
1201824
304 SS Port Thread Inserts, two 1/4 in.-28 White PTFE, PP, SS Nalgene 38-430
Each for $87.00
Add to Cart
 
13-311-027
View Documents
Cole-Parmer™
1201827
Two 1/4 in.-28 ports White PTFE, PP Nalgene 38-430
Each for $73.50
Add to Cart
 
13-311-041
View Documents
Cole-Parmer™
1201841
Four 1/4 in.-28 ports White PTFE, PP Wako 38 mm
Each for $73.25
Add to Cart
 
13-311-038
View Documents
Cole-Parmer™
1201838
304 SS Port Thread Inserts, four 1/4 in.-28 White PTFE, PP, SS Wako 38 mm
Each for $87.00
Add to Cart
 
13-311-000
View Documents
Cole-Parmer™
1201800
304 SS Port Thread Inserts, two 1/4 in.-28 Blue/white PTFE, PP, SS GL45
Each for $84.00
Add to Cart
 
13-311-053
View Documents
Cole-Parmer™
1201853
Four 1/4 in.-28 ports Blue/white PTFE, PP GL32
Each for $66.25
Add to Cart
 
13-311-048
View Documents
Cole-Parmer™
1201848
304 SS Port Thread Inserts, two 1/4 in.-28 Blue/white PTFE, PP, SS GL32
Each for $87.00
Add to Cart
 
13-311-055
View Documents
Cole-Parmer™
1201855
Three 1/4 in.-28 ports Blue/white PTFE, PP GL25
Each for $56.75
Add to Cart
 
13-311-012
View Documents
Cole-Parmer™
1201812
304 SS Port Thread Inserts, two 1/4 in.-28 Black/white PTFE, PP, SS GL38
Each for $89.00
Add to Cart
 
13-311-001
View Documents
Cole-Parmer™
1201801
304 SS Port Thread Inserts, three 1/4 in.-28 Blue/white PTFE, PP, SS GL45
Each for $108.00
Add to Cart
 
13-311-039
View Documents
Cole-Parmer™
1201839
Two 1/4 in.-28 ports White PTFE, PP Wako 38 mm
Each for $75.00
Add to Cart
 
13-311-017
View Documents
Cole-Parmer™
1201817
Four 1/4 in.-28 ports Black/white PTFE, PP GL38
Each for $74.50
Add to Cart
 
13-311-026
View Documents
Cole-Parmer™
1201826
304 SS Port Thread Inserts, four 1/4 in.-28 White PTFE, PP, SS Nalgene 38-430
Each for $92.00
Add to Cart
 
13-311-036
View Documents
Cole-Parmer™
1201836
304 SS Port Thread Inserts, two 1/4 in.-28 White PTFE, PP, SS Wako 38 mm
Each for $79.00
Add to Cart
 
13-311-016
View Documents
Cole-Parmer™
1201816
Three 1/4 in.-28 ports Black/white PTFE, PP GL38
Each for $73.50
Add to Cart
 
13-311-040
View Documents
Cole-Parmer™
1201840
Three 1/4 in.-28 ports White PTFE, PP Wako 38 mm
Each for $66.00
Add to Cart
 
13-311-029
View Documents
Cole-Parmer™
1201829
Four 1/4 in.-28 ports White PTFE, PP Nalgene 38-430
Each for $64.50
Add to Cart
 
13-311-054
View Documents
Cole-Parmer™
1201854
Two 1/4 in.-28 ports Blue/white PTFE, PP GL25
Each for $58.75
Add to Cart
 
13-311-002
View Documents
Cole-Parmer™
1201802
304 SS Port Thread Inserts, four 1/4 in.-28 Blue/white PTFE, PP, SS GL45
Each for $124.00
Add to Cart
 
13-311-014
View Documents
Cole-Parmer™
1201814
304 SS Port Thread Inserts, four 1/4 in.-28 Black/white PTFE, PP, SS GL38
Each for $98.00
Add to Cart
 
Description

Description

Use with semi-rigid tubing equal to or less than 1/8 in (3.2 mm) OD

Warranty and Services

None

Specifications

Specifications

Solvent Safety
Solvent-Compatible Containers
1/4 in.-28 UNF(F)
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.