Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cole-Parmer™ VapLock™ Inlet Check Valves for Solvent Management
VapLock™ Inlet Check Valves for Solvent Management maintain pressure equilibrium within the bottle for safe solvent handling
$73.25 - $111.00
Specifications
Type | Inlet Check Valve; 0.1-0.25 psi crack pressure |
---|---|
For Use With (Application) | Solvent Safety |
For Use With (Equipment) | VapLock Solvent Containment Systems |
Includes | 1 Ea. |
Thread Size (English) | 1/4 in.-28 UNF(M) |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
13-311-263
|
Cole-Parmer™
1202061 |
PTFE, FFKM |
Each for $111.00
|
|
|||||
13-311-254
|
Cole-Parmer™
1202052 |
PTFE, Viton |
Each for $73.25
|
|
|||||
13-311-255
|
Cole-Parmer™
1202053 |
PTFE, EPDM |
Each for $76.00
|
|
|||||
Description
- Minimize evaporation and relieve pressure to improve safety
- Allow air into supply container to displace vacuum and keep solvent flowing
Warranty and Services
None
Specifications
Inlet Check Valve; 0.1-0.25 psi crack pressure | |
VapLock Solvent Containment Systems | |
1/4 in.-28 UNF(M) |
Solvent Safety | |
1 Ea. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title