Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cole-Parmer™ VapLock™ Adapters for Semi-Rigid Tubing, UNF(M)
VapLock™ Adapters for Semi-Rigid Tubing, UNF(M) are designed for secure liquid-tight connections
$19.70 - $142.10
Specifications
For Use With (Application) | Solvent Safety |
---|---|
For Use With (Equipment) | VapLock Solvent Containment Systems |
Catalog Number | Mfr. No. | Size | Thread Size (English) | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Thread Size (English) | Material | Price | Quantity & Availability | |||
13-311-203
|
Cole-Parmer™
1202001 |
4 mm OD Compression x UNF(M) | 5/16 in.-24 UNF(M) | Brown / White PEEK / PCTFE |
Each for $27.25
|
|
|||
13-311-204
|
Cole-Parmer™
1202002 |
3/16 in. OD Compression x UNF(M) | 5/16 in.-24 UNF(M) | White / Blue POM / ETFE |
Each for $19.70
|
|
|||
13-311-205
|
Cole-Parmer™
1202003 |
1/16 in. OD Compression x UNF(M) | 1/4 in.-28 UNF(M) | Green / Blue PP / ETFE |
Pack of 10 for $93.40
|
|
|||
13-311-206
|
Cole-Parmer™
1202004 |
2 mm OD Compression x UNF(M) | 1/4 in.-28 UNF(M) | Red / Green PP / ETFE |
Pack of 10 for $116.20
|
|
|||
13-311-207
|
Cole-Parmer™
1202005 |
2.2 mm OD Compression x UNF(M) | 1/4 in.-28 UNF(M) | Black / White PP / PTFE |
Pack of 10 for $65.40
|
|
|||
13-311-208
|
Cole-Parmer™
1202006 |
2.5 mm OD Compression x UNF(M) | 1/4 in.-28 UNF(M) | Orange / White PP / ETFE |
Pack of 10 for $118.30
|
|
|||
13-311-209
|
Cole-Parmer™
1202007 |
3 mm OD Compression x UNF(M) | 1/4 in.-28 UNF(M) | Yellow / Orange PP / ETFE |
Pack of 10 for $142.10
|
|
|||
13-311-210
|
Cole-Parmer™
1202008 |
1/8 in. OD Compression x UNF(M) | 1/4 in.-28 UNF(M) | Blue / Yellow PP / ETFE |
Pack of 10 for $92.30
|
|
|||
Description
- Color-coded for easy identification
- Features two-piece nut and ferrule system
Warranty and Services
None
Specifications
Solvent Safety | |
VapLock Solvent Containment Systems |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title