Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Uranine K, TCI America™
Click to view available options
Quantity:
25 g
Specifications
Specifications
Quantity | 25 g |
Formula Weight | 408.49 |
Physical Form | Crystalline Powder |
Chemical Name or Material | Uranine K |
Safety and Handling
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction