Learn More
Bioworld Newborn Calf Serum (<14 days of age), 100 mL
Supplier: Bioworld 306110761
Newborn calf serum will support a wide variety of established cell lines and primary cultures which might otherwise require fetal bovine serum. Empirical testing is the most reliable method for determining usefulness for a particular line.SpecificationsSterile-filteredMycoplasma-tested and virus-screened Ships on dry ice. U.S. or N.Z. Origin (dependant on availiability)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.