Learn More
DuPont™ Tychem™ 9000 Hood with Standard Visor
Click to view available options
Tychem™ 9000 Hood with Standard Visor
Description
DuPont™ Tychem™ 9000 is strong and durable for consistent and lasting protection.
- NFPA 1994 Class 2 and NFPA 1992 certified
- Provides at least 30 minutes of protection against over 280 chemical challenges
- Used by many in the chemical handling and petrochemical manufacturing
Specifications
Specifications
Size | Universal |
Material | Multi-layer Laminate |
Color | Yellow |
Closure Type | Hook and Loop |
Seam Type | Taped |
Resistance | Chemical |
Style | Standard Visor |