Learn More
Tosoh Bioscience Reproductive Hormone Assays
Hormone assay tests and accessories
Supplier: Tosoh Bioscience 020355
This item is not returnable.
View return policy
Description
- Single, unitized cups use a dry reagent format that requires no pre-mixing, no pre-measuring, no on-board refrigeration and no waste
- The AIA test cups and trays are labeled with code and lot number for automated scheduling and inventory management on AIA-2000, AIA-1800 and AIA-900 sorter models
- ST AIA-PACK reagents provide a faster turnaround time due to a 10 minute incubation protocol
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.
Specifications
Prolactin | |
Prolactin Calibrator Set | |
4 x 1 mL |