Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Tolcapone

COMT inhibitor; also inhibits transthyretin aggregation

$127.70 - $530.00

Specifications

Chemical Name or Material (3,4-Dihydroxy-5-nitrophenyl)(4-methylphenyl)methanone
CAS 134308-13-7
Synonym Tasmar, Ro 40-7592
Target Catechol O-Methyltransferase (COMT) Inhibitors
Molecular Formula C14H11NO5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
586410
View Documents
Tocris Bioscience™
5864/10
10 mg
Each for $127.70
Add to Cart
 
586450
View Documents
Tocris Bioscience™
5864/50
50 mg
Each for $530.00
Add to Cart
 
Description

Description

COMT inhibitor. Inhibits both brain and peripheral COMT. Also binds transthyretin (TTR) with high affinity (Kd1 and Kd2 values are 21 and 58 nM, respectively). Inhibits TTR aggregation in human plasma and prevents TTR-induced cytotoxicity in vitro. Stabilizes TTR in mice and humans in vivo. Orally bioavailable.
Specifications

Specifications

(3,4-Dihydroxy-5-nitrophenyl)(4-methylphenyl)methanone
Tasmar, Ro 40-7592
C14H11NO5
>99%
134308-13-7
Catechol O-Methyltransferase (COMT) Inhibitors
273.24
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.