Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TGX 221

Potent and selective PI 3-kinase β inhibitor

$412.96 - $1730.72

Specifications

Chemical Name or Material 7-Methyl-2-(4-morpholinyl)-9-[1-(phenylamino)ethyl]-4H-pyrido[1,2-a]pyrimidin-4-one
CAS 663619-89-4
Target PI 3-Kinase Inhibitors
Molecular Formula C21H24N4O2
Formula Weight 364.44
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
583210
View Documents
Tocris Bioscience™
5832/10
10 mg
Each for $412.96
Add to Cart
 
583250
View Documents
Tocris Bioscience™
5832/50
50 mg
Each for $1,730.72
Add to Cart
 
Description

Description

Potent and selective PI 3-kinase β inhibitor (IC50 values are 0.007, 0.1, 3.5 and 5 μM for the β, δ, γ and α isoforms, respectively). Shows >1,000-fold selectively for PI 3-kinase β over a range of other kinases. Inhibits thrombus formation in animal models.
Specifications

Specifications

7-Methyl-2-(4-morpholinyl)-9-[1-(phenylamino)ethyl]-4H-pyrido[1,2-a]pyrimidin-4-one
PI 3-Kinase Inhibitors
364.44
663619-89-4
C21H24N4O2
>98%
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.