Learn More
Leica Biosystems STA-ON TISSUE ADHESIVE
Supplier: Leica Biosystems 3803107
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Sta-On is a liquid adhesive that is easy to use and convenient. 10mL are added to each liter of pre-warmed water in a tissue floatation bath. Diluted Sta-On can also be used to coat slides.