Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 9009

Rev-Erbα/β agonist

$243.60 - $999.92

Specifications

Chemical Name or Material Ethyl 3-[[[(4-chlorophenyl)methyl][(5-nitro-2-thienyl)methyl]amino]methyl]-1-pyrrolidinecarboxylate
CAS 1379686-30-2
Target Nuclear Receptor Agonists
Molecular Formula C20H24ClN3O4S
Formula Weight 437.94
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
58-551-0
View Documents
Tocris Bioscience™
5855/10
10 mg
Each for $243.60
Add to Cart
 
58-555-0
View Documents
Tocris Bioscience™
5855/50
50 mg
Each for $999.92
Add to Cart
 
Description

Description

Rev-Erbα/β agonist (EC50 = 710 nM; Kd = 800 nM). Alters circadian locomotor activity and core clock hypothalamic gene expression in mice. Induces weight loss in diet-induced obese mice. Brain penetrant.
Specifications

Specifications

Ethyl 3-[[[(4-chlorophenyl)methyl][(5-nitro-2-thienyl)methyl]amino]methyl]-1-pyrrolidinecarboxylate
Nuclear Receptor Agonists
437.94
1379686-30-2
C20H24ClN3O4S
>98%
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.