Learn More
Wilmad Labglass SPRAYER TLC COMP 125ML
Supplier: Wilmad Labglass LG4590100
Sprayer, TLC Reagent, Complete. Capacity: 125mL; Joint Size: [ST] 24/40. For spraying chromagenic reagents when using thin layer or paper chromatography. Complete unit consists of spray head, flask and LG-1004-108 plastic clamp.