Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SouthernBiotech™ Normal Mouse Sera
Processed to prevent minimal hemolysis and filtered to 0.2μm
Supplier: SouthernBiotech™ 005001
Description
Blocking sera; Control sera; Serum additive in dilution buffers
Specifications
RUO | |
2° to 8°C | |
Mouse Serum | |
Blocking Sera, control Sera, Serum additive in dilution Buffers | |
8 |
Colorless to Light Brown | |
Adult Non-Swiss Albino Mouse | |
5 mL | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for Diagnostic or Therapeutic Use.
Spot an opportunity for improvement?
Provide Content Correction