Learn More
Camag Scientific Smartcut Plate Cutter To Cut Tlc/Hptlc Glass Plates Up To 20 X 20 Cm
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Camag Scientific 022.4300
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
The Smartcut Allows Cutting Of Tlc And Hptlc Glass Plates In Defined Pieces Of Any Format. The Sensitive Layer Is Not Destroyed And The Resulting Cut Has Smooth Edges.