Learn More
Uline Silica Gel Desiccant 5 Gallon Pail
Supplier: Uline S23158
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
25 LB reusable silica gel desiccant beads in 5 GL pail adsorb moisture and control humidity via physical adsorption to protect contents during storage and transport.