Learn More
Epredia™ Shandon™ Digital Scale, Replacement bow
Get accurate autopsy and pathology weight measurements with the Shandon™ Digital Scale.
$285.00
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity & Availability | |||||
10-601-9
|
Epredia™
106019 |
Digital Scale Replacement Bow |
Each for $285.00
|
|
1000232
|
Epredia™
1000232 |
Digital Scale Replacement Pan | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
1330
|
Epredia™
1330 |
Digital Scale (Order Stand Separately) | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
- Displays readings with bright, easy-to-read LCD
- Measures up to 19.99kg
- Includes ring, bow and pan with drainage holes
- Battery operated
Compatible with:
Replacement parts may also be used with Shandon™ Analog Scale (Cat. No. 1320).