Learn More
Abraxis Saxitoxin Certified Standard | 20ug/ml | 0.5ml
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Abraxis 300590
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
The Saxitoxins Certified Standard undergoes rigorous analysis for identity and purity, including NMR and UPLC-MS/MS. The intended uses are for the calibration of equipment or a measurement procedure; establishing metrological traceability; method validation; or quality control of a measurement or measurement procedure.