Learn More
Polar Tech Industries Re-Freez-R-Brix 7oz 4 x 4 x ", RB 8 (42/CS)
SDP
Supplier:  Polar Tech Industries RB 8
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
Polar Tech's foam 'ReFreezRBrix' is our top selling ice pack brand companywide. These ice packs offer consistent shape, durability, longevity, reliability, and they are great for using in packouts that have limited flexibility with space. The foam block's density and thickness makes the product less susceptible to thawing, making them last about 10% longer than gel packs. If you want a working machine, these ice packs are the ones for you. They will do their job and do it well.
Catalog No. NC1589850
Explore available promotions