Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Euramco Ramfan™ Air-Driven Blowers: Adapter/Reducer
Supplier: Euramco EC0301
Specifications
Duct adapter/reducer, 8 in. | |
For use with 12 in. AFi air-driven blower; Also suitable for EFi series electric blowers |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title