Learn More
Thermo Scientific RADIAL EMT TORCH
Supplier: Thermo Scientific 842312051741
Radial Torch (EMT); ICP quartz torch for use with iCAP 6000 Series Radial instruments; using the EMT Torch holder.