Learn More
Associates of Cape Cod Inc PYROTELL LAL 5ML 5VIALS/PACK
SDP
Supplier:  Associates of Cape Cod Inc G51255
Encompass
PYROTELL GEL CLOT LAL, 0.125 EU/mL SENSITIVITY, 5mL/VIAL, 5 VIALS/PACK
Catalog No. NC9001884