Learn More
iQ Biosciences Purified Cynomolgus Monkey NK Cells, 1M cells/vial
Supplier: iQ Biosciences IQBMN1N1
High-quality, high-viability (> 90%) NK cells purified from cynomolgus (cyno) monkey (or macaque) PBMCs by magnetic selection. Highly enriched (typically > 80%) post-selection purity achieved via magnetic isolation. Used for a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.The following donor requests can be accommodated if inventory is available: - Single or multiple donors for a particular product (SKU). Indicate number of vials from each individual donor - Donor-matched PBMCs
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.