Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ ProSpecT™ Cryptosporidium Microplate Assay
Rapidly detect Crytosporidium specific antigen from human fecal samples.
$2313.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Description | ProSpecT Cryptosporidium Microplate Assay |
Detectable Analytes | Cryptosporidium Antigen |
Includes | Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card |
Catalog Number | Mfr. No. | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Quantity | Content And Storage | Price | Quantity & Availability | |||
R2454096
|
Thermo Scientific™
R2454096 |
ProSpecT Cryptosporidium Microplate Assay | 96 Wells/Kit | 2°C to 8°C |
Pack of 96 for $2,313.00
|
|
|||
Description
Rapidly detect Crytosporidium specific antigens (CSA) in fresh or preserved fecal specimens using Thermo Scientific™ ProSpecT™ Cryptosporidium Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Crytosporidium. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.- Excellent clinical performance
- Cary Blair transport option which saves a dilution step
- Results within 100 minutes
- Room temperature incubation
- True colorless negatives
ProSpecT Cryptosporidium provides same-day antigen detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C | |
ProSpecT Cryptosporidium Microplate Assay | |
Cryptosporidium Antigen | |
Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title