Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Iron-Total Liquid Reagents
Quantify iron levels in serum. Use with Pointe Scientific™ Iron Standard Reagent (Catalog No. 23-666-312)
$94.00 - $355.00
Specifications
Certifications/Compliance | CE Marked |
---|---|
Formulation | Iron Buffer R1: 220mM Hydroxylamine Hydrochloride in Acetate Buffer, pH 4.5 with Surfactant, Iron color R2: 3.6mM Ferrozine in Hydroxylamine Hydrochloride |
For Use With (Application) | Quantitative determination of Iron in Serum |
Form | Liquid |
Includes: Buffer Reagent: Hydroxylamine hydrochloride 220mM in acetate buffer (pH 4.5); surfactant
Color Reagent: Ferrozine 16.7mM in hydroxylamine hydrochloride
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity & Availability | ||||
23-666-317
|
Pointe Scientific
I7505120 |
1 x 120mL Buffer, 1 x 5mL Color | Total Iron Reagent Set 1 |
Each for $94.00
|
|
||||
23-666-318
|
Pointe Scientific
I7505500 |
1 x 500mL Buffer, 1 x 15mL Color | Total Iron Reagent Set 2 |
Each for $355.00
|
|
||||
23-666-316
|
Pointe Scientific
HI704300 |
5 x 50mL R1, 1 x 50mL R2 | Iron (Ferrozine) Reagent | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Method: Ferrozine, Endpoint
- Format: Liquid (ready to use)
- Wavelength: 560nm
- Linearity: 500ug/dL
- Expected Values: 60 to 150ug/dL
For the quantitative determination of iron in serum
Specifications
CE Marked | |
Iron Buffer R1: 220mM Hydroxylamine Hydrochloride in Acetate Buffer, pH 4.5 with Surfactant, Iron color R2: 3.6mM Ferrozine in Hydroxylamine Hydrochloride | |
Quantitative determination of Iron in Serum | |
Liquid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title