Learn More
Biocare Medical, LLC PeroxAbolish - 500 ml
Supplier: Biocare Medical, LLC PXA969M
PeroxAbolish is a novel formula that is a non-hydrogen peroxide blocking reagent that is safe and extremely gentle on tissues and cells. It is highly effective in quenching endogenous peroxidase and can be used on formalin-fixed paraffin-embedded tissues, cell culture, blood smears, cell preparations and frozen sections. When using paraffin-embedded formalin-fixed tissues, PeroxAbolish is extremely effective after antigen retrieval methods. It can also be used on an automated staining system.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.