Learn More
CPC Scientific H-gamma-Glu-Abu-Gly-OH (trifluoroacetate salt) 1GR
Supplier: CPC Scientific OARP001B
Acidic peptide of the lens. Carba analog of reduced glutathione. Ophthalmic acid could be used as a biomarker in oxidative stress.ONE-LETTER SEQUENCE: gamma-E-Abu-GMOLECULAR FORMULA: C11H19N3O6MOLECULAR WEIGHT:289.29STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [495-27-2], RESEARCH AREA: MiscellaneousREFERENCES: S.G.Waley, Biochem. J., 68, 189 (1958); T.Soga et al., J. Biol. Chem., 281, 16768 (2006)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.