Learn More
AbD Serotec MOUSE SERUM,PRESRTV FREE,100ML
Supplier: AbD Serotec C11SDZ
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Mouse Serum:Preservative FreeProduct Form; Whole serum - liquidPreparation; Normal mouse serum, 0.2um filtered. Prepared from barrier mice which are screened for infectious agents.Preservative Stabilisers; None presentStorage; Store at -20oC only. This product should be stored undiluted.Storage in frost-free freezers is not recommended. Avoid repeated freezing and thawing as this may denature the product. Should this product contain a precipitate we recommend microcentrifugation before use
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.