Learn More
Fisherbrand™ Micro Stir Bars
Micro Color-Coded Stirring Bar For stirring test tubes, micro-beakers, etc.
$54.25 - $149.00
Catalog Number | Quantity | Color | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Color | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
S29164
|
24 | Yellow, Blue, Red | Multiple sizes |
Pack of 24 for $149.00
|
|
||||
S29165
|
12 | Red | 0.31 x 0.07 in. (8 x 1.7 mm) |
Pack of 12 for $58.25
|
|
||||
S29166
|
12 | Yellow | 0.31 x 0.07 in. (8 x 1.7 mm) |
Pack of 12 for $54.25
|
|
||||
S29167
|
12 | Blue | 0.31 x 0.07 in. (8 x 1.7 mm) |
Pack of 12 for $58.25
|
|
||||
S29169
|
12 | Yellow | 0.19 x 0.08 in. (5 x 2 mm) |
Pack of 12 for $60.75
|
|
||||
S29168
|
12 | Red | 0.19 x 0.08 in. (5 x 2 mm) |
Pack of 12 for $60.75
|
|
||||
S29172
|
12 | Yellow | 0.27 x 0.09 in. (7 x 2.2 mm) |
Pack of 12 for $62.25
|
|
||||
S29173
|
12 | Blue | 0.27 x 0.08 in. (7 x 2 mm) |
Pack of 12 for $60.75
|
|
||||
S29174
|
12 | Red | 0.39 x 0.126 in. (10 x 3.2 mm) |
Pack of 12 for $67.25
|
|
||||
S29175
|
12 | Yellow | 0.39 x 0.13 in. (10 x 3.3 mm) |
Pack of 12 for $67.25
|
|
||||
S29177
|
12 | Blue | 0.39 x 0.126 in. (10 x 3.2 mm) |
Pack of 12 for $67.25
|
|
||||
S29179
|
12 | Red | 0.5 x 0.128 in. (12.7 x 3.25 mm) |
Pack of 12 for $67.25
|
|
||||
S29180
|
12 | Yellow | 0.5 x 0.128 in. (12.7 x 3.25 mm) |
Pack of 12 for $67.25
|
|
||||
S29181
|
12 | Blue | 0.5 x 0.124 in. (12.7 x 3.14 mm) |
Pack of 12 for $67.25
|
|
||||
S29171
|
12 | Red | 0.27 x 0.09 in. (7 x 2.2 mm) | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||
Description
- Octagonal shape
- Resists corrosion, heat, and cold
- Available in blue, red or yellow
- PTFE
Specifications
PTFE | |
Octagon |