Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Isobutyl Ketone, Exceeds A.C.S. Specifications, HPLC Grade, 99%, Spectrum™ Chemical
SDP

Catalog No. 18611704
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 108-10-1

Specifications

CAS Max % 1
Appearance Clear
Percent Purity 0.99
Grade High Purity
Quantity 1 L
Packaging Amber Glass Bottle
Residue after Evaporation 5 ppm
Titratable Acid 2 μeq/g
Water 0.0008

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.