Learn More
Bellco Glass, Inc. Metal stone sparger 36l for flat or round bottom omni vessels. Sparger inserts through stainless steal headplate fitting. 4 1/2"wide x 24" long 316 l stainless steel ra 32 finish or better Stainless steel frit
Supplier: Bellco Glass, Inc. 196489436
Metal stone sparger 36l for flat or round bottom omni vessels. Sparger inserts through stainless steal headplate fitting. 4 1/2"wide x 24" long 316 l stainless steel ra 32 finish or better Stainless steel frit