Learn More
Ace Glass, Inc. MERCURY RESERVOIR MERCURY RESERVOIR
Supplier: Ace Glass, Inc. 781842
Mercury Reservoir with a #25 Ace-Thred for the ACE-Burlitch system. The ACE-Burlitch inert atmosphere system is designed for maximum convenience and versatility in the handling of air-sensitive compounds in conjunction with ACE no-air labware, or suitiably adapted conventional glassware.