Learn More
Eisco™ Measuring Cylinder with Protective Collar, Sure Trace Certification
Graduated Cylinder, ASTM, Class A. Protective Collar, Hexagonal Base, Blue Graduations. Made of Borosilicate 3.3 Glass.
Supplier: Eisco™ CH0341BBLST
Description
- Measuring Cylinder made of high quality, heavy duty, Borosilicate 3.3 glass
- Class A Tolerance, as per ISO/ASTM standards
- Easy to read, blue printed graduations and specifications.
- Cylinder features a hexagonal base for maximum stability and is fitted with a removable, plastic protective collar
- Superior chemical durability and heat tolerance.
- Designed for use in professional laboratories, but is an ideal choice for science classrooms and household use
Specifications
Graduated Cylinder | |
±0.17 mL | |
25 mL |
Class A | |
Borosilicate Glass (3.3) | |
0.5 mL |