Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Maxima H Minus Reverse Transcriptase (200 U/μL) PROMO

Maxima H Minus Reverse Transcriptase (RT) was developed through in vitro evolution of M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity but lacks RNase H activity due to mutation in RNase H domain of M-MuLV RT.

Supplier:  Thermo Scientific™ EP0752

 View more versions of this product

Catalog No. FEREP0752PM


Edge
Explore available promotions
Add to Cart

Description

Description

Thermo Scientific Maxima H Minus Reverse Transcriptase (RT) was developed through in vitro evolution of M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity but lacks RNase H activity due to mutation in RNase H domain of M-MuLV RT. The engineered enzyme features dramatically improved thermostability, 50X higher processivity, robustness, and increased synthesis rate compared to wild type M-MuLV RT.

The eliminated RNase H activity enables the enzyme to produce very long RNA transcripts up to 20 kb. Due to its high thermostability, the enzyme maintains full activity during the entire reverse transcription reaction and generates high yields of cDNA. The reaction temperature can be increased to 65°C for efficient transcription of RNA regions with a high secondary structure or to improve specificity using gene specific primers. The extremely high processivity of Maxima H Minus enzyme results in increased resistance to common reaction inhibitors, such as guanidine, formamide, and ethanol.

Features of Maxima H Minus Reverse Transcriptase include:

  • Thermostable–90% active after incubation at 50°C for 60 minutes in a reaction mixture
  • Active up to 65°C
  • High yields of full-length cDNA up to 20 kb
  • High sensitivity–reproducible cDNA synthesis from a wide range of starting total RNA amounts (1 pg to 5 μg)
  • Efficient–complete cDNA synthesis in 15 to 30 minutes
  • Increased resistance to common reaction inhibitors
  • Incorporates modified nucleotides

Applications

  • First strand cDNA synthesis for RT-PCR and RT-qPCR
  • Reverse transcription at elevated temperatures to reduce effects of secondary structure
  • Synthesis of cDNA for cloning and expression
  • Generation of labeled cDNA probes for microarrays
  • Analysis of RNA by primer extension

Related products

  • Maxima H Minus Reverse Transcriptase (Cat. No. EP0752)
  • Maxima H Minus First Strand cDNA Synthesis Kit (Cat. No. K1652)
  • Maxima H Minus First Strand cDNA Synthesis Kit, with dsDNase (Cat. No. K1681)
  • Maxima H Minus cDNA Synthesis Master Mix (Cat. No. M1661)
  • Maxima H Minus cDNA Synthesis Master Mix, with dsDNase (Cat. No. M1681)
Specifications

Specifications

Maxima H Minus Reverse Transcriptase, 1 x 10,000 units (200 U/μL), 5X RT Buffer , Store at -20°C.
High
Reverse Transcription
10,000 Units
Reduced
200 U/μL
Separate components
Up to 20 kb
Stand-alone Enzyme
15 to 30 min.
50°C to 55°C
Maxima H Minus
Dry Ice
First-Strand cDNA
Reverse Transcription
RNA
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

For Research Use Only. Not for use in diagnostic procedures.