Learn More
Sigma Aldrich Fine Chemicals Biosciences Luperox A98, Benzoyl peroxide, 94-36-0, MFCD00003071, 500g
Supplier: Sigma Aldrich Fine Chemicals Biosciences 179981500G
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Linear Formula (C6H5CO)2O2, ≥98%, Molecular Weight 242.23, reagent grade, mp 105 °C (lit.), Synonym: Benzoyl peroxide, Dibenzoyl peroxide; Benzoyl peroxide participates in the covalent insitu functionalization of carbon nanotubes.
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)